Learn More
8-(tert-Butyldimethylsilyloxy) 8-Hydroxy Efavirenz, Technical Grade, TRC
Specifications
Specifications
| Chemical Name or Material | 8-(tert-Butyldimethylsilyloxy) 8-Hydroxy Efavirenz, Technical Grade |
| CAS | 1027042-31-4 |
| Molecular Formula | C20H23ClF3NO3Si |
| Quantity | 5 mg |
| Synonym | 8-(tert-Butyldimethylsilyloxy)-6-chloro-4-(cyclopropylethynyl)-1,4-dihydro-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one, 6-Chloro-4-(2-cyclopropylethynyl)-8-[[(1,1-dimethylethyl)dimethylsilyl]oxy]-1,4-dihydro-4-(trifluoromethyl)-2H-3,1-benzoxazin-2-one |
| InChI Formula | InChI=1S/C20H23ClF3NO3Si/c1-18(2,3)29(4,5)28-15-11-13(21)10-14-16(15)25-17(26)27-19(14,20(22,23)24)9-8-12-6-7-12/h10-12H,6-7H2,1-5H3,(H,25,26) |
| SMILES | CC(C)(C)[Si](C)(C)Oc1cc(Cl)cc2c1NC(=O)OC2(C#CC3CC3)C(F)(F)F |
| IUPAC Name | 8-[tert-butyl(dimethyl)silyl]oxy-6-chloro-4-(2-cyclopropylethynyl)-4-(trifluoromethyl)-1H-3,1-benzoxazin-2-one |
| Molecular Weight (g/mol) | 445.94 |
| Formula Weight | 445.1088 |