missing translation for 'onlineSavingsMsg'
Learn More

8-Methoxypsoralen, 99%, MP Biomedicals™

Product Code. 11427521 Shop All MP Biomedicals Products
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
This item is not returnable. View return policy

Product Code. 11427521

Brand: MP Biomedicals 0210329610

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 298-81-7
Molecular Formula C12H8O4
Molecular Weight (g/mol) 216.19
MDL Number MFCD00005009
InChI Key QXKHYNVANLEOEG-UHFFFAOYSA-N
Synonym methoxsalen, 8-methoxypsoralen, xanthotoxin, meladinine, ammoidin, oxsoralen, meloxine, oxypsoralen, xanthotoxine, meladinin
PubChem CID 4114
ChEBI CHEBI:18358
IUPAC Name 9-methoxy-7H-furo[3,2-g]chromen-7-one
SMILES COC1=C2OC=CC2=CC2=C1OC(=O)C=C2

Specifications

Color White to Cream
Assay Percent Range 0.99
Quantity 10 g
Formula Weight 216.2
Physical Form Powder
Chemical Name or Material 8-Methoxypsoralen
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.