missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline Sulfate Monohydrate, Sigma-Aldrich™

Product Code. 15829970
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15829970 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15829970 Supplier Sigma Aldrich Supplier No. 8145280100
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 207386-91-2
Melting Point 182°C
MDL Number MFCD00149610
Quantity 100 g
Synonym 8-Hydroxyquinoline sulfate monohydrate, Quinosol, 8-Hydroxyquinoline sulfate monohydrate
Molecular Weight (g/mol) 388.39 (Anhydrous Basis)
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.