missing translation for 'onlineSavingsMsg'
Learn More

8-Hydroxyquinoline-5-sulfonic Acid Monohydrate, Sigma-Aldrich™

Product Code. 15879920
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15879920 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications
Product Code. 15879920 Supplier Sigma-Aldrich Supplier No. 8144360100
 Restricted Item
Estimated Shipment: 11-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
MQ200 Graded - For Research and Non-regulated Industrial Applications

For synthesis

TRUSTED_SUSTAINABILITY

Specifications

CAS 283158-18-9
Melting Point 322°C
MDL Number MFCD00149482
Quantity 100 g
Synonym 8-Hydroxyquinoline-5-sulfonic acid monohydrate, 8-Hydroxyquinoline-5-sulfonic acid monohydrate
Molecular Weight (g/mol) 243.24
Grade Research
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.