Learn More
8-Hydroxyjulolidine, 97%
Product Code. 11413278
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
Product Code. 11413278
Brand: Thermo Scientific Alfa Aesar A17692.03
Request Bulk or Custom Format
CAS: 41175-50-2 | C12H15NO | 189.258 g/mol
8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
8-Hydroxyjulolidine is used as an intermediate in the preparation of aminocoumarins via microwave-accelerated Pechmann reaction. It is also employed as a starting material for coumarin C-ribosides.
Solubility
Soluble in water (partly), ethanol, chloroform, methanol, and acetone.
Notes
Store away from oxidizing agents and strong bases. Keep the container tightly closed in a cool, dry place.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 41175-50-2
Molecular Formula C12H15NO
MDL Number MFCD00006918
InChI Key FOFUWJNBAQJABO-UHFFFAOYSA-N
PubChem CID 170474
Specifications
Melting Point 132°C to 135°C
Quantity 1 g
Beilstein 1530423
Synonym 8-hydroxyjulolidine, 1,2,3,5,6,7-hexahydropyrido 3,2,1-ij quinolin-8-ol, 2,3,6,7-tetrahydro-1h,5h-benzo ij quinolizin-8-ol, 1h,5h-benzo ij quinolizin-8-ol, 2,3,6,7-tetrahydro, 2,3,6,7-tetrahydro-1h,5h-pyrido 3,2,1-ij quinolin-8-ol, 8-hydroxy julolidine, timtec-bb sbb006615, 1-azatricyclo 7.3.1.0^ 5,13 trideca-5 13 ,6,8-trien-6-ol
Solubility Information Soluble in water (partly),ethanol,chloroform,methanol,and acetone.
SMILES C1CC2=C3C(=C(C=C2)O)CCCN3C1
Molecular Weight (g/mol) 189.258
Formula Weight 189.26
Percent Purity 97%
Chemical Name or Material 8-Hydroxyjulolidine