CAS RN 41175-50-2
IUPAC Name:
1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8-trien-6-ol
Synonyms:
1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8-trien-6-ol
Molecular Weight (g/mol):
189.26
Molecular Formula:
C12H15NO
InChi Key:
FOFUWJNBAQJABO-UHFFFAOYSA-N
SMILES:
OC1=CC=C2CCCN3CCCC1=C23
1
–
2
of
2
results
1
8-Hydroxyjulolidine, 97%
CAS: 41175-50-2 Molecular Formula: C12H15NO MDL Number: MFCD00006918 InChI Key: FOFUWJNBAQJABO-UHFFFAOYSA-N PubChem CID: 170474 IUPAC Name: 1-azatricyclo[7.3.1.0⁵,¹³]trideca-5(13),6,8-trien-6-ol