missing translation for 'onlineSavingsMsg'
Learn More

8-Chlorotheophylline, TRC

Product Code. 30007132
Change view
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30007132 10 g 10g
17964825 25 g 25g
17940435 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30007132 Supplier Toronto Research Chemicals Supplier No. TRCC41144010G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 85-18-7
Molecular Formula C7 H7 Cl N4 O2
Molecular Weight (g/mol) 214.61
Synonym 1H-Purine-2,6-dione, 8-chloro-3,9-dihydro-1,3-dimethyl-, 1H-Purine-2,6-dione, 8-chloro-3,7-dihydro-1,3-dimethyl- (9CI), Theophylline, 8-chloro- (6CI,7CI,8CI), 8-Chloro-3,9-dihydro-1,3-dimethyl-1H-purine-2,6-dione, 1,3-Dimethyl-8-chloroxanthine, 8-Chloro-1,3-dimethylxanthine, 8-Chlorotheophylline, NSC 6113
IUPAC Name 8-chloro-1,3-dimethyl-7H-purine-2,6-dione
SMILES CN1C(=O)N(C)c2nc(Cl)[nH]c2C1=O

Specifications

Quantity 10 g
InChI Formula InChI=1S/C7H7ClN4O2/c1-11-4-3(9-6(8)10-4)5(13)12(2)7(11)14/h1-2H3,(H,9,10)
Formula Weight 214.0258
Chemical Name or Material 8-Chlorotheophylline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.