Learn More
8-Chloro-5-isoquinolinesulfonic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 8-Chloro-5-isoquinolinesulfonic Acid |
| CAS | 1246816-17-0 |
| Molecular Formula | C9H6ClNO3S |
| Quantity | 250 mg |
| Synonym | 8-Chloro-5-isoquinoline Sulfonic Acid |
| InChI Formula | InChI=1S/C9H6ClNO3S/c10-8-1-2-9(15(12,13)14)6-3-4-11-5-7(6)8/h1-5H,(H,12,13,14) |
| SMILES | OS(=O)(=O)c1ccc(Cl)c2cnccc12 |
| IUPAC Name | 8-chloroisoquinoline-5-sulfonic acid |
| Molecular Weight (g/mol) | 243.67 |
| Formula Weight | 242.9757 |