Learn More
8-Chloro-5,6-dihydro-11H-benzo[5,6]cyclohepta[1,2-b]pyridin-11-one, TRC
Specifications
Specifications
| Chemical Name or Material | 8-Chloro-5,6-dihydro-11H-benzo[5.6]cyclohepta[1,2-b]pyridin-11-one |
| CAS | 31251-41-9 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C14 H10 Cl N O |
| Quantity | 250 mg |
| Synonym | 8-Chloro-5,6-dihydro-11H-benzo[5.6]cyclohepta[1,2-b]pyridin-11-one, Loratadine Imp. B (EP) |
| InChI Formula | InChI=1S/C14H10ClNO/c15-11-5-6-12-10(8-11)4-3-9-2-1-7-16-13(9)14(12)17/h1-2,5-8H,3-4H2 |
| SMILES | Clc1ccc2C(=O)c3ncccc3CCc2c1 |
| IUPAC Name | 8-chloro-5,6-dihydrobenzo[1,2]cyclohepta[2,4-b]pyridin-11-one |
| Molecular Weight (g/mol) | 243.69 |
| Show More |
Product Title
Haben Sie Verbesserungsvorschläge?