missing translation for 'onlineSavingsMsg'
Learn More

(8-Carboxyoctyl)triphenylphosphonium Bromide, TRC

Product Code. 30366206
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Packungsgröße:
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Quantity unitSize
30366206 1 g 1g
30365646 100 mg -
30353858 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 30366206 Lieferant Toronto Research Chemicals Lieferanten-Nr. TRCC1789051G
 Restricted Item
Estimated Shipment: 27-08-2026
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88462-46-8
Molecular Formula C27H32BrO2P
Molecular Weight (g/mol) 499.42
Synonym 9-(Bromotriphenylphosphoranyl)nonanoic Acid, (8-Carboxyoctyl)triphenyl-phosphonium Bromide
IUPAC Name 8-carboxyoctyl(triphenyl)phosphanium;bromide
SMILES OC(=O)CCCCCCCCP(Br)(c1ccccc1)(c2ccccc2)c3ccccc3

Specifications

Quantity 1 g
InChI Formula InChI=1S/C27H31O2P.BrH/c28-27(29)22-14-3-1-2-4-15-23-30(24-16-8-5-9-17-24,25-18-10-6-11-19-25)26-20-12-7-13-21-26;/h5-13,16-21H,1-4,14-15,22-23H2;1H
Formula Weight 498.1323
Chemical Name or Material (8-Carboxyoctyl)triphenylphosphonium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.