Learn More
(8-Carboxyoctyl)triphenylphosphonium Bromide, TRC
Product Code. 30365646
Change view
Click to view available options
Quantity:
50 mg
100 mg
1 g
Unit Size:
1g
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365646 100 mg -
30353858 50 mg 50mg
30366206 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30365646 Supplier Toronto Research Chemicals Supplier No. TRCC178905100MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 88462-46-8
Molecular Formula C27H32BrO2P
Molecular Weight (g/mol) 499.42
Synonym 9-(Bromotriphenylphosphoranyl)nonanoic Acid, (8-Carboxyoctyl)triphenyl-phosphonium Bromide
IUPAC Name 8-carboxyoctyl(triphenyl)phosphanium;bromide
SMILES OC(=O)CCCCCCCCP(Br)(c1ccccc1)(c2ccccc2)c3ccccc3
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C27H31O2P.BrH/c28-27(29)22-14-3-1-2-4-15-23-30(24-16-8-5-9-17-24,25-18-10-6-11-19-25)26-20-12-7-13-21-26;/h5-13,16-21H,1-4,14-15,22-23H2;1H
Formula Weight 498.1323
Chemical Name or Material (8-Carboxyoctyl)triphenylphosphonium Bromide