missing translation for 'onlineSavingsMsg'
Learn More

8-Bromo-1-octanol, 95%

Product Code. 11416580
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11416580

Brand: Thermo Scientific Alfa Aesar H27628.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 50816-19-8 | C8H17BrO | 209.127 g/mol

It is used to produce 8-phenylselanyl-octan-1-ol by reaction with diphenyl-diselane. The reaction occurs with reagent sodium borohydride and solvent ethanol at ambient temperature. The yield is about 96%. 8-Bromo-1-octanol was used in the synthesis of (E)-10-hydroxy-2-decenoic acid (royal jelly acid)1. It was also used in the synthesis of (Z)-14-methyl-9-pentadecenoic acid.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used to produce 8-phenylselanyl-octan-1-ol by reaction with diphenyl-diselane. The reaction occurs with reagent sodium borohydride and solvent ethanol at ambient temperature. The yield is about 96%. 8-Bromo-1-octanol was used in the synthesis of (E)-10-hydroxy-2-decenoic acid (royal jelly acid)1. It was also used in the synthesis of (Z)-14-methyl-9-pentadecenoic acid.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents. Refrigerated (4°C).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 50816-19-8
Molecular Formula C8H17BrO
Molecular Weight (g/mol) 209.127
MDL Number MFCD00010388
InChI Key GMXIEASXPUEOTG-UHFFFAOYSA-N
Synonym 8-bromo-1-octanol, 1-octanol, 8-bromo, 8-bromooctanol, octamethylene bromohydrin, 8-bromo octanol, 1-bromooctan-8-ol, 8-bromo-1-actanol, 8-bromooctane-1-ol, 1-bromo-octan-8-ol, 8-bromo-octan-1-ol
PubChem CID 162607
IUPAC Name 8-bromooctan-1-ol
SMILES C(CCCCBr)CCCO

Specifications

Density 1.22
Boiling Point 79°C to 80°C (0.07 mmHg)
Flash Point >110°C (230°F)
Linear Formula Br(CH2)8OH
Refractive Index 1.481
Quantity 25 g
Beilstein 1900837
Formula Weight 209.13
Percent Purity 95%
Chemical Name or Material 8-Bromo-1-octanol

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.