missing translation for 'onlineSavingsMsg'
Learn More

8-Aminonaphthalene-1-sulfonic acid, 98%

Product Code. 11308566
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11308566 25 g 25g
11318566 100 g 100g
11328566 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11308566 Supplier Thermo Scientific Alfa Aesar Supplier No. L17616.14
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 82-75-7 | C10H9NO3S | 223.246 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 82-75-7
Molecular Formula C10H9NO3S
Molecular Weight (g/mol) 223.246
MDL Number MFCD00035730
InChI Key CYJJLCDCWVZEDZ-UHFFFAOYSA-N
Synonym peri acid, 8-amino-1-naphthalenesulfonic acid, 1-naphthylamine-8-sulfonic acid, 1-naphthalenesulfonic acid, 8-amino, 8-aminonaphthalene-1-sulphonic acid, naphthylaminemonosulfonic acid s, 1-amino-8-naphthalene sulfonate, 1-aminonaphthalene-8-sulfonic acid, schollkopf's acid van, schollkopf's acid
PubChem CID 6722
IUPAC Name 8-aminonaphthalene-1-sulfonic acid
SMILES C1=CC2=C(C(=C1)N)C(=CC=C2)S(=O)(=O)O

Specifications

Melting Point >300°C
Quantity 25 g
Beilstein 983230
Merck Index 14,6405
Formula Weight 223.25
Percent Purity 98%
Chemical Name or Material 8-Aminonaphthalene-1-sulfonic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.