missing translation for 'onlineSavingsMsg'
Learn More

7-p-Toluenesulfonylhydrazide 3Beta,4Beta-Dihydroxy-chol-5-ene-24-carboxylic Acid Ethyl Ester, TRC

Product Code. 30455807
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455807 10 mg 10mg
30453269 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30455807 Supplier Toronto Research Chemicals Supplier No. TRCT53591010MG
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-p-Toluenesulfonylhydrazide 3beta,4beta-Dihydroxy-chol-5-ene-24-carboxylic Acid Ethyl Ester
Analyte or Component Names 7-p-Toluenesulfonylhydrazide 3beta,4beta-Dihydroxy-chol-5-ene-24-carboxylic Acid Ethyl Ester
Shipping Condition Room Temperature
Molecular Formula C34H50N2O6S
InChI Formula InChI=1S/C34H50N2O6S/c1-6-42-30(38)9-7-8-22(3)24-14-15-25-31-26(16-18-33(24,25)4)34(5)19-17-29(37)32(39)27(34)20-28(31)35-36-43(40,41)23-12-10-21(2)11-13-23/h10-13,20,22,24-26,29,31-32,36-37,39H,6-9,14-19H2,1-5H3/b35-28-/t22-,24-,25+,26+,29+,31+,32-,33-,34-/m1/s1
Solution Type Neat
SMILES [H][C@@]1([C@]2(C)C([C@@H](O)[C@@H](O)CC2)=C/3)[C@@]([C@@](CC[C@]4([H])[C@H](C)CCCC(OCC)=O)([H])[C@]4(C)CC1)([H])C3=N/NS(C5=CC=C(C)C=C5)(=O)=O
IUPAC Name ethyl (R)-5-((3S,4R,8S,9S,10R,13R,14S,17R,E)-3,4-dihydroxy-10,13-dimethyl-7-(2-tosylhydrazineylidene)-2,3,4,7,8,9,10,11,12,13,14,15,16,17-tetradecahydro-1H-cyclopenta[a]phenanthren-17-yl)hexanoate
Industry Type Pharmaceutical
Molecular Weight (g/mol) 614.84
Quantity 10 mg
Formula Weight 614.34
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.