missing translation for 'onlineSavingsMsg'
Learn More
Learn More
7-Methylxanthine, 98%
Click to view available options
Quantity:
50 mg
250 mg
Unit Size:
250mg
50mg
Description
It is used to metabolite theophylline and caffeine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
It is used to metabolite theophylline and caffeine.
Solubility
Soluble in dimethyl sulfoxide. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
It is used to metabolite theophylline and caffeine.
Solubility
Soluble in dimethyl sulfoxide. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 50 mg |
| Solubility Information | Soluble in dimethyl sulfoxide. Insoluble in water. |
| IUPAC Name | 7-methyl-3H-purine-2,6-dione |
| Formula Weight | 166.14 |
| Percent Purity | 98% |
| Chemical Name or Material | 7-Methylxanthine |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction