accessibility menu, dialog, popup

UserName

CAS RN 552-62-5

H3CNNHNONHO

CAS RN 552-62-5

IUPAC Name: 7-methyl-2,3,6,7-tetrahydro-1H-purine-2,6-dione
Synonyms: 7-methylxanthine
Molecular Weight (g/mol): 166.14
Molecular Formula: C6H6N4O2
InChi Key: PFWLFWPASULGAN-UHFFFAOYSA-N
SMILES: CN1C=NC2=C1C(=O)NC(=O)N2
Quantity 
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (2)
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)