missing translation for 'onlineSavingsMsg'
Learn More

7-Methyl-3-methyluric Acid-d3, TRC

Product Code. 30472806
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30472806 2.5 mg 2.5mg
30478000 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30472806 Supplier Toronto Research Chemicals Supplier No. TRCM3309002.5MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 383160-11-0
Molecular Formula C7 D3 H5 N4 O3
Molecular Weight (g/mol) 199.18
IUPAC Name 7-methyl-3-(trideuteriomethyl)-9H-purine-2,6,8-trione
SMILES [2H]C([2H])([2H])N1C(=O)NC(=O)C2=C1NC(=O)N2C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C7H8N4O3/c1-10-3-4(8-6(10)13)11(2)7(14)9-5(3)12/h1-2H3,(H,8,13)(H,9,12,14)/i2D3
Formula Weight 199.0785
Chemical Name or Material 7-Methyl-3-(methyl-d3)uric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.