CAS RN 383160-11-0
CAS RN 383160-11-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Methyl-3-methyluric Acid-d3, TRC
CAS: 383160-11-0 Molecular Formula: C7 D3 H5 N4 O3 Molecular Weight (g/mol): 199.18 IUPAC Name: 7-methyl-3-(trideuteriomethyl)-9H-purine-2,6,8-trione SMILES: [2H]C([2H])([2H])N1C(=O)NC(=O)C2=C1NC(=O)N2C