Learn More
7-Hydroxyflavone, 98%
Product Code. 11428457
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11428457 1 g 1g
11438457 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11428457 Supplier Thermo Scientific Alfa Aesar Supplier No. A18538.03
 Restricted Item
In Stock
Add to basket
CAS: 6665-86-7 | C15H10O3 | 238.242 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6665-86-7
Molecular Formula C15H10O3
Molecular Weight (g/mol) 238.242
MDL Number MFCD00006835
InChI Key MQGPSCMMNJKMHQ-UHFFFAOYSA-N
Synonym 7-hydroxyflavone, flavone, 7-hydroxy, 7-hydroxy-2-phenyl-4-benzopyrone, 4h-1-benzopyran-4-one, 7-hydroxy-2-phenyl, 7-hydroxy-2-phenyl-4h-chromen-4-one, 4h-1-benzopyran-4-one, 7-hydroxy, unii-ze72458e4l, 7-hydroxy-2-phenyl-chromen-4-one, 7-hydroxy-2-phenyl-4h-1-benzopyran-4-one, 7-hydroxyflavone, 11
PubChem CID 5281894
ChEBI CHEBI:2268
IUPAC Name 7-hydroxy-2-phenylchromen-4-one
SMILES C1=CC=C(C=C1)C2=CC(=O)C3=C(O2)C=C(C=C3)O
Specifications
Melting Point 245°C to 247°C
Quantity 1 g
Beilstein 194089
Formula Weight 238.25
Percent Purity 98%
Chemical Name or Material 7-Hydroxyflavone