Learn More
7-Epi Lincomycin Hydrochloride Salt, TRC
Product Code. 30134138
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30134138 5 mg 5mg
30137251 10 mg 10mg
30008233 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30134138 Supplier Toronto Research Chemicals Supplier No. TRCE5885505MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 26389-84-4
Molecular Formula C18H34N2O6S • HCl
Molecular Weight (g/mol) 406.543646
Synonym Methyl 6,8-dideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1- thio-L-threo-α-D-galacto-octopyranoside Hydrochloride, trans-α-Methyl 6,8-dideoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)- 1-thio-L-threo-D-galacto-octopyranoside Hydrochloride, (7S)-Lincomycin Hydrochloride, 7-Deoxy-7(S)-hydroxylincomycin Hydrochloride, Epilincomycin Hydrochloride, U 20244E Hydrochloride;
SMILES O[C@H]1[C@@H]([C@H](NC(C2C[C@@H](CCC)CN2C)=O)[C@](O)([H])C)O[C@H](SC)C(O)C1O.Cl
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C18H34N2O6S.ClH/c1-5-6-10-7-11(20(3)8-10)17(25)19-12(9(2)21)16-14(23)13(22)15(24)18(26-16)27-4;/h9-16,18,21-24H,5-8H2,1-4H3,(H,19,25);1H
Formula Weight 442.1904
Percent Purity >95%
Chemical Name or Material 7-Epi Lincomycin Hydrochloride Salt
Grade HPLC