CAS RN 26389-84-4
CAS RN 26389-84-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
7-Epi Lincomycin Hydrochloride Salt, TRC
CAS: 26389-84-4 Molecular Formula: C18H34N2O6S • HCl Molecular Weight (g/mol): 406.543646 Synonym: Methyl 6,8-dideoxy-6-[[[(2S,4R)-1-methyl-4-propyl-2-pyrrolidinyl]carbonyl]amino]-1- thio-L-threo-α-D-galacto-octopyranoside Hydrochloride,trans-α-Methyl 6,8-dideoxy-6-(1-methyl-4-propyl-L-2-pyrrolidinecarboxamido)- 1-thio-L-threo-D-galacto-octopyranoside Hydrochloride,(7S)-Lincomycin Hydrochloride,7-Deoxy-7(S)-hydroxylincomycin Hydrochloride,Epilincomycin Hydrochloride,U 20244E Hydrochloride; SMILES: O[C@H]1[C@@H]([C@H](NC(C2C[C@@H](CCC)CN2C)=O)[C@](O)([H])C)O[C@H](SC)C(O)C1O.Cl