Learn More
Tocris Bioscience™ 7-Chlorokynurenic acid sodium salt
Description
7-Chlorokynurenic acid sodium salt is a sodium salt of 7-Chlorokynurenic acid (Cat.No. 0237), an NMDA receptor antagonist acting at the glycine site. Potent competitive inhibitor of L-glutamate transport into synaptic vesicles.
Specifications
Specifications
| Quantity | 50 mg |
| Product Type | 7-Chlorokynurenic acid sodium salt |
| Purity | >98% |
| Molecular Weight (g/mol) | 245.59 |
| Inhibitors | 7-Chlorokynurenic acid sodium salt |
For Research Use Only