Learn More
7-Chloro-6-fluoro-1-(4-fluorophenyl)-1,4-dihydro-4-oxo-3-quinolinecarboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 7-Chloro-6-fluoro-1-(4-fluorophenyl)-4-oxo-1,4-dihydroquinoline-3-carboxylic Acid |
| CAS | 98105-79-4 |
| Molecular Formula | C16 H8 Cl F2 N O3 |
| Quantity | 100 mg |
| Synonym | 7-Chloro-6-fluoro-1-(4-fluorophenyl)-1,4-dihydro-4-oxo-3-quinolinecarboxylic acid, Difloxacin Impurity G |
| InChI Formula | InChI=1S/C16H8ClF2NO3/c17-12-6-14-10(5-13(12)19)15(21)11(16(22)23)7-20(14)9-3-1-8(18)2-4-9/h1-7H,(H,22,23) |
| SMILES | OC(=O)C1=CN(c2ccc(F)cc2)c3cc(Cl)c(F)cc3C1=O |
| IUPAC Name | 7-chloro-6-fluoro-1-(4-fluorophenyl)-4-oxoquinoline-3-carboxylic acid |
| Molecular Weight (g/mol) | 335.69 |
| Formula Weight | 335.0161 |