Learn More
7-(But-2-yn-1-yl)-3-methyl-8-(methylamino)-1-((4-methylquinazolin-2-yl)methyl)-1H-purine-2,6(3H,7H)-dione, TRC
Specifications
Specifications
| Chemical Name or Material | 7-(But-2-yn-1-yl)-3-methyl-8-(methylamino)-1-((4-methylquinazolin-2-yl)methyl)-1H-purine-2,6(3H,7H)-dione |
| CAS | 2468656-63-3 |
| Molecular Formula | C21 H21 N7 O2 |
| Quantity | 50 mg |
| Synonym | Linagliptin Impurity |
| InChI Formula | InChI=1S/C21H21N7O2/c1-5-6-11-27-17-18(25-20(27)22-3)26(4)21(30)28(19(17)29)12-16-23-13(2)14-9-7-8-10-15(14)24-16/h7-10H,11-12H2,1-4H3,(H,22,25) |
| SMILES | CNc1nc2N(C)C(=O)N(Cc3nc(C)c4ccccc4n3)C(=O)c2n1CC#CC |
| IUPAC Name | 7-but-2-ynyl-3-methyl-8-(methylamino)-1-[(4-methylquinazolin-2-yl)methyl]purine-2,6-dione |
| Molecular Weight (g/mol) | 403.437 |
| Formula Weight | 403.176 |