missing translation for 'onlineSavingsMsg'
Learn More

7-Bromoheptanoic Acid, TRC

Product Code. 30347062
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347062 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30347062 Supplier Toronto Research Chemicals Supplier No. TRCB687890100MG
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Bromoheptanoic Acid
CAS 30515-28-7
Molecular Formula C7H13BrO2
Quantity 100 mg
Synonym zeta-Bromo-enanthic Acid, 7-Bromoheptanoic Acid, ω-Bromoheptanoic Acid
InChI Formula InChI=1S/C7H13BrO2/c8-6-4-2-1-3-5-7(9)10/h1-6H2,(H,9,10)
SMILES OC(=O)CCCCCCBr
IUPAC Name 7-bromoheptanoic acid
Molecular Weight (g/mol) 209.08
Formula Weight 208.0099
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.