Learn More
7-Bromoheptanoic acid, 97%
Product Code. 15392042 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15392042 25 g 25g
15362042 1 g 1g
15372042 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15392042 Supplier Thermo Scientific Acros Supplier No. 453260250
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 30515-28-7 | C7H13BrO2 | 209.08 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30515-28-7
Molecular Formula C7H13BrO2
Molecular Weight (g/mol) 209.08
InChI Key JLPQXFFMVVPIRW-UHFFFAOYSA-N
Synonym 7-bromo-heptanoic acid, heptanoic acid, 7-bromo, 7-bromoenanthic acid, omega-bromoheptanoic acid, 7-bromoheptanoicacid, 5-bromopentylacetate;, 7-bromo heptanoic acid, acmc-1aezo, ksc226m8n
PubChem CID 121723
IUPAC Name 7-bromoheptanoic acid
SMILES C(CCCBr)CCC(=O)O
Specifications
Melting Point 29.0°C to 32.0°C
Boiling Point 121.0°C to 125.0°C (5.0 mmHg)
Infrared Spectrum Authentic
Quantity 25 g
Formula Weight 209.08
Percent Purity 97%
Chemical Name or Material 7-Bromoheptanoic acid
compliance-icons
Product Identifier
  • 7-Bromoheptanoic acid, 97%
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.