missing translation for 'onlineSavingsMsg'
Learn More

7-Bromo-DL-tryptophan (contains up to 20% inorganics), TRC

Product Code. 30348002
Change view
Click to view available options
Quantity:
2.5 g
Unit Size:
2.50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30348002 2.5 g 2.50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30348002 Supplier Toronto Research Chemicals Supplier No. TRCB6790252.5G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 7-Bromo-DL-tryptophan
CAS 852391-45-8
Molecular Formula C11 H11 Br N2 O2
Quantity 2.5 g
InChI Formula InChI=1S/C11H11BrN2O2/c12-8-3-1-2-7-6(5-14-10(7)8)4-9(13)11(15)16/h1-3,5,9,14H,4,13H2,(H,15,16)
SMILES NC(Cc1c[nH]c2c(Br)cccc12)C(=O)O
IUPAC Name 2-amino-3-(7-bromo-1H-indol-3-yl)propanoic acid
Molecular Weight (g/mol) 283.121
Formula Weight 282
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.