missing translation for 'onlineSavingsMsg'
Learn More

7,7,8,8-Tetracyanoquinodimethane, 98%

Product Code. p-7080985
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11409692 5 g 5g
11419692 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11409692 Supplier Thermo Scientific Alfa Aesar Supplier No. A10779.06
Add to basket
This item is not returnable. View return policy

CAS: 1518-16-7 | C12H4N4 | 204.192 g/mol

7,7,8,8-Tetracyanoquinodimethane is an electron-acceptor molecule used to form charge-transfer superconductors. It is an effective catalyst used for the ?-chlorination of carboxylic acids using chlorosulfonic acid; the presence of TCNQ suppresses competing free-radical chlorination.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7,7,8,8-Tetracyanoquinodimethane is an electron-acceptor molecule used to form charge-transfer superconductors. It is an effective catalyst used for the ɑ-chlorination of carboxylic acids using chlorosulfonic acid; the presence of TCNQ suppresses competing free-radical chlorination.

Solubility
Insoluble in water.

Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1518-16-7
Molecular Formula C12H4N4
Molecular Weight (g/mol) 204.192
MDL Number MFCD00011664
InChI Key PCCVSPMFGIFTHU-UHFFFAOYSA-N
Synonym 7,7,8,8-tetracyanoquinodimethane, tcnq, tetracyanoquinodimethane, tetracyanoquinodimethan, 2,2'-cyclohexa-2,5-diene-1,4-diylidene dimalononitrile, tetracyano-p-quinodimethane, tetracyanoquinodimethan e, quinodimethan, tetracyano, propanedinitrile, 2,2'-2,5-cyclohexadiene-1,4-diylidene bis, 7,7,8,8-tetracyano-p-quinodimethane
PubChem CID 73697
ChEBI CHEBI:52445
IUPAC Name 2-[4-(dicyanomethylidene)cyclohexa-2,5-dien-1-ylidene]propanedinitrile
SMILES C1=CC(=C(C#N)C#N)C=CC1=C(C#N)C#N

Specifications

Melting Point ∼288°C (decomposition)
Quantity 5 g
UN Number UN3439
Beilstein 611604
Solubility Information Insoluble in water.
Formula Weight 204.19
Percent Purity 98%
Chemical Name or Material 7,7,8,8-Tetracyanoquinodimethane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.