missing translation for 'onlineSavingsMsg'
Learn More

7-(2-Chloroethyl)theophylline, 97%, Thermo Scientific Chemicals

Product Code. 15466729
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15466729 5 g 5g
15456729 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15466729 Supplier Thermo Scientific Alfa Aesar Supplier No. J65111.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

An adenosine receptor antagonist

An adenosine receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5878-61-5
Molecular Formula C9H11ClN4O2
Molecular Weight (g/mol) 242.663
MDL Number MFCD00005760
InChI Key QCIARNIKNKKHFH-UHFFFAOYSA-N
Synonym 7-2-chloroethyl theophylline, benaphyllin, eupnophile, 7-chloroethyl theophylline, theophylline, 7-2-chloroethyl, beta-chloroethyltheophylline, unii-y7202un6b7, 7-beta-chlorethyl theophylline, 7-chloroethyltheophylline, 1h-purine-2,6-dione, 7-2-chloroethyl-3,7-dihydro-1,3-dimethyl
PubChem CID 1882
IUPAC Name 7-(2-chloroethyl)-1,3-dimethylpurine-2,6-dione
SMILES CN1C2=C(C(=O)N(C1=O)C)N(C=N2)CCCl

Specifications

Melting Point 122°C to 130°C
Color White
Quantity 5 g
Solubility Information Soluble in DMSO; Moderately soluble in ethanol.
Formula Weight 242.66
Percent Purity 97%
Physical Form Powder
Chemical Name or Material 7-(2-Chloroethyl)theophylline

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.