missing translation for 'onlineSavingsMsg'
Learn More

6Beta-Methoxy-3,5-cycloandrost-15-en-17-one, TRC

Product Code. 30465381
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30465381 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30465381 Supplier Toronto Research Chemicals Supplier No. TRCM2610551MG
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6Beta-Methoxy-3,5-cycloandrost-15-en-17-one
CAS 1309594-70-4
Molecular Formula C20H28O2
Quantity 1 mg
InChI Formula InChI=1S/C20H28O2/c1-18-8-7-15-13(14(18)4-5-16(18)21)10-17(22-3)20-11-12(20)6-9-19(15,20)2/h4-5,12-15,17H,6-11H2,1-3H3/t12?,13-,14-,15-,17+,18-,19+,20?/m0/s1
SMILES CO[C@@H]1C[C@@H]2[C@H](CC[C@@]3(C)[C@H]2C=CC3=O)[C@@]4(C)CCC5CC145
Molecular Weight (g/mol) 300.44
Formula Weight 300.2089
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.