Learn More
6alpha-Hydroxy Androstenedione, TRC
Specifications
Specifications
| Chemical Name or Material | 6alpha-Hydroxy Androstenedione |
| CAS | 24704-84-5 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C19 H26 O3 |
| Quantity | 25 mg |
| Synonym | Androst-4-ene-3,17-dione, 6-hydroxy-, (6alpha)-, Androst-4-ene-3,17-dione, 6alpha-hydroxy- (8CI), (6alpha)-6-Hydroxyandrost-4-ene-3,17-dione, 6alpha-Hydroxy-4-androstene-3,17-dione, 6alpha-Hydroxyandrostenedione, Androst-4-en-6alpha-ol-3,17-dione |
| InChI Formula | InChI=1S/C19H26O3/c1-18-7-5-11(20)9-15(18)16(21)10-12-13-3-4-17(22)19(13,2)8-6-14(12)18/h9,12-14,16,21H,3-8,10H2,1-2H3/t12-,13-,14-,16-,18+,19-/m0/s1 |
| SMILES | C[C@]12CC[C@H]3[C@@H](C[C@H](O)C4=CC(=O)CC[C@]34C)[C@@H]1CCC2=O |
| IUPAC Name | (6S,8R,9S,10R,13S,14S)-6-hydroxy-10,13-dimethyl-2,6,7,8,9,11,12,14,15,16-decahydro-1H-cyclopenta[a]phenanthrene-3,17-dione |
| Molecular Weight (g/mol) | 302.41 |
| Show More |