Learn More
MedChemExpress 6-ROX
Click to view available options
:
5 mg
Specifications
Specifications
| Quantity | 5 mg |
| Product Type | Dye Reagents |
| Molecular Weight (g/mol) | 534.6 g/mol |
| Solubility | DMSO : 25 mg/mL (46.76 mM; Need ultrasonic) |
| Content And Storage | 4°C, protect from light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (protect from light) |
| Shipping Condition | Room temperature |
| Target | HBV |
| CAS | 194785-18-7 |
| Synonym | 6-Carboxy-X-rhodamine |