Learn More
Thermo Scientific™ 6-Place Fixed-Angle Highconic™ Rotor Adapters
Product Code. 10676373 Shop All Thermo Scientific Products
Change view
Click to view available options
Tube Type:
16 mL Tube
Conical
Conical Falcon
Microtube
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Tube Type Unit Size
10676373 Conical Each
10107881 Microtube Each
10085060 16 mL Tube Each
10318612 Conical Falcon Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10676373 Supplier Thermo Scientific™ Supplier No. 75003102
  • ONLINE EXCLUSIVE PRICE
    94.50 EUR
    91.20 EUR / Pack of 1
    Save 3.30 EUR
    3% Off
In Stock
Requires
50mL conical adapter 75-007-556 requires rubber pads (supplied with buckets). Remove pads when using all other size adapters.
Use these rotor adapters to connect with round or conical-bottom centrifuge tubes and microtubes tubes up to 50mL.
Use these 6-Place Fixed-Angle Highconic™ Rotor Adapters to connect with centrifuge tubes up to 50mL. The adapters accommodate round or conical-bottom tubes and microtubes and work with HiGhConic™ fixed angle rotors.
  • The adapters accommodate round- or conical-bottom tubes and microtubes.
  • Rotor adaptors work with HiGhConic™ fixed angle rotors
  • Fits tube sizes up to 50mL
Order Info
Adapters are sold individually; order correct quantity to equip rotor.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Tube Type Conical
Capacity 50 mL
For Use With (Equipment) HiGhConic™ fixed angle rotor
Tube Size 50 mL