Learn More
6-Nitro-1-pyrenol-d8, TRC
Product Code. 30468820
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30468820 10 mg 10mg
30475407 1 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30468820 Supplier Toronto Research Chemicals Supplier No. TRCN51998710MG
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 2469736-26-1
Molecular Formula C16HD8NO3
Molecular Weight (g/mol) 271.3
Synonym 1-Nitro-6-hydroxypyrene-d8, 1-Nitropyren-6-ol-d8, 6-Hydroxy-1-nitropyrene-d8
IUPAC Name 6-nitropyren-2,3,4,5,7,8,9,10-d8-1-ol
SMILES OC1=C([2H])C([2H])=C2C3=C1C([2H])=C([2H])C4=C([2H])C([2H])=C([N+]([O-])=O)C(C([2H])=C2[2H])=C43
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C16H9NO3/c18-14-8-4-10-1-5-11-13(17(19)20)7-3-9-2-6-12(14)16(10)15(9)11/h1-8,18H/i1D,2D,3D,4D,5D,6D,7D,8D
Formula Weight 271.11
Chemical Name or Material 6-Nitro-1-pyrenol-d8