missing translation for 'onlineSavingsMsg'
Learn More

6-Methylquinoline, TRC

Product Code. 30464770
Change view
Click to view available options
Quantity:
10 g
100 g
Unit Size:
100g
10g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30464770 100 g 100g
30479728 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30464770 Supplier Toronto Research Chemicals Supplier No. TRCM328005100G
 Restricted Item
Estimated Shipment: 30-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91-62-3
Molecular Formula C10 H9 N
Molecular Weight (g/mol) 143.185
Synonym 6-Methylquinoline, NSC 4152
IUPAC Name 6-methylquinoline
SMILES Cc1ccc2ncccc2c1

Specifications

Quantity 100 g
InChI Formula InChI=1S/C10H9N/c1-8-4-5-10-9(7-8)3-2-6-11-10/h2-7H,1H3
Formula Weight 143.0735
Chemical Name or Material 6-Methylquinoline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.