missing translation for 'onlineSavingsMsg'
Learn More

6-Methylquinoline, 98%

Product Code. 12131220 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 g
Packaging:
Glass bottle
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
12131220 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 12131220 Supplier Thermo Scientific Acros Supplier No. 295140250

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 91-62-3 | C10H9N | 143.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 91-62-3
Density 1.0600g/mL
Boiling Point 256.0°C to 260.0°C
Flash Point >113°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H9N
Refractive Index 1.6125 to 1.6145
MDL Number MFCD00006804
Quantity 25 g
Specific Gravity 1.06
Synonym quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
Solubility Information Solubility in water: .. Other solubilities: soluble in ethanol
InChI Key LUYISICIYVKBTA-UHFFFAOYSA-N
SMILES CC1=CC=C2N=CC=CC2=C1
IUPAC Name 6-methylquinoline
Molecular Weight (g/mol) 143.19
PubChem CID 7059
Formula Weight 143.19
Percent Purity 98%
Chemical Name or Material 6-Methylquinoline
Show More Show Less
compliance-icons
Product Identifier
  • 6-Methylquinoline, 98%
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H315-Causes skin irritation.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P362-Take off contaminated clothing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.