Learn More
6-Methylquinoline, 98%
Product Code. 11414967
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11414967 100 g 100g
11354639 5 g 5g
11404967 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11414967 Supplier Thermo Scientific Alfa Aesar Supplier No. B21892.22
 Restricted Item
Estimated Shipment: 09-09-2026
Add to basket
CAS: 91-62-3 | C10H9N | 143.19 g/mol
6-Methylquinoline can be used as primary carbon source in culture of Pseudomonas putida QP1. 6-Methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
6-Methylquinoline can be used as primary carbon source in culture of Pseudomonas putida QP1. 6-Methylquinoline was used in the synthesis of fluorescent halide-sensitive quinolinium dyes and fluorescent probes for determination of chloride in biological systems.
Solubility
Not miscible or difficult to mix with water.
Notes
Light Sensitive, store in dark. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 91-62-3
Molecular Formula C10H9N
Molecular Weight (g/mol) 143.19
MDL Number MFCD00006804
InChI Key LUYISICIYVKBTA-UHFFFAOYSA-N
Synonym quinoline, 6-methyl, p-toluquinoline, 6-methyl-quinoline, tolliquinoline, p, ccris 407, fema no. 2744, quinoline, 6-methyl-8ci,9ci, dsstox_cid_887, dsstox_rid_75844, dsstox_gsid_20887
PubChem CID 7059
IUPAC Name 6-methylquinoline
SMILES CC1=CC=C2N=CC=CC2=C1
Specifications
Melting Point -22°C
Density 1.066
Boiling Point 257°C to 259°C
Flash Point >110°C (230°F)
Refractive Index 1.615
Quantity 100 g
Beilstein 110336
Sensitivity Light sensitive
Solubility Information Not miscible or difficult to mix with water.
Formula Weight 143.19
Percent Purity 98%
Chemical Name or Material 6-Methylquinoline