Learn More
6-Methyl-4-(trifluoromethyl)pyridin-2(1H)-one, 95%, BLD Pharm™
Spezifikation
Spezifikation
| CAS | 22123-19-9 |
| Molecular Formula | C7H6F3NO |
| MDL Number | MFCD00042479 |
| Quantity | 1 g |
| SMILES | O=C1C=C(C(F)(F)F)C=C(C)N1 |
| Molecular Weight (g/mol) | 177.12 |
| Percent Purity | 95% |
| Chemical Name or Material | 6-Methyl-4-(trifluoromethyl)pyridin-2(1H)-one |