Learn More
6-Methoxyindole-2-carboxylic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Methoxyindole-2-carboxylic Acid |
| CAS | 16732-73-3 |
| Molecular Formula | C10H9NO3 |
| Quantity | 10 g |
| Synonym | 6-Methoxy-1H-indole-2-carboxylic Acid, 6-Methoxyindole-2-carboxylic Acid, NSC 27988 |
| InChI Formula | InChI=1S/C10H9NO3/c1-14-7-3-2-6-4-9(10(12)13)11-8(6)5-7/h2-5,11H,1H3,(H,12,13) |
| SMILES | COc1ccc2cc([nH]c2c1)C(=O)O |
| IUPAC Name | 6-methoxy-1H-indole-2-carboxylic acid |
| Molecular Weight (g/mol) | 191.18 |
| Formula Weight | 191.0582 |