missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxychrysene, TRC

Product Code. 30040663
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30040663 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30040663 Supplier Toronto Research Chemicals Supplier No. TRCH9053501MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

CAS 37515-51-8
Molecular Formula C18 H12 O
Quantity 1 mg
Synonym 6-Chrysenol (6CI, 7CI, 9CI, ACI), 6-Hydroxychrysene
InChI Formula InChI=1S/C18H12O/c19-18-11-17-13-6-2-1-5-12(13)9-10-15(17)14-7-3-4-8-16(14)18/h1-11,19H
SMILES Oc1cc2c3ccccc3ccc2c4ccccc14
IUPAC Name chrysen-6-ol
Molecular Weight (g/mol) 244.29
Formula Weight 244.0888
Chemical Name or Material 6-Hydroxychrysene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.