missing translation for 'onlineSavingsMsg'
Learn More

6-Hydroxy Bentazon, TRC

Product Code. 30132773
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30132773 5 mg 5mg
17905035 10 mg 10mg
17973435 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30132773 Supplier Toronto Research Chemicals Supplier No. TRCH8065005MG
 Restricted Item
Estimated Shipment: 27-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 60374-42-7
Molecular Formula C10 H12 N2 O4 S
Molecular Weight (g/mol) 256.28
Synonym 1H-2,1,3-Benzothiadiazin-4(3H)-one, 6-hydroxy-3-(1-methylethyl)-, 2,2-dioxide, 6-Hydroxybentazon, 6-Hydroxybentazone, 1H-2,1,3-Benzothiadiazin-4(3H)-one, 6-hydroxy-3-(1-methylethyl)-, 2,2-dioxide, 6-Hydroxybentazon, 6-Hydroxybentazone
IUPAC Name 6-hydroxy-2,2-dioxo-3-propan-2-yl-1H-2λ^{6},1,3-benzothiadiazin-4-one
SMILES CC(C)N1C(=O)c2cc(O)ccc2NS1(=O)=O

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H12N2O4S/c1-6(2)12-10(14)8-5-7(13)3-4-9(8)11-17(12,15)16/h3-6,11,13H,1-2H3
Formula Weight 256.0518
Percent Purity >95%
Chemical Name or Material 6-Hydroxy bentazone
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.