Learn More
6-Hydroxy-2-naphthoic acid, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 16712-64-4 |
| Molecular Formula | C11H8O3 |
| MDL Number | MFCD00060070 |
| Quantity | 100 g |
| SMILES | O=C(O)C1=CC=C2C=C(O)C=CC2=C1 |
| Molecular Weight (g/mol) | 188.18 |
| Percent Purity | 98% |
| Chemical Name or Material | 6-Hydroxy-2-naphthoic acid |