missing translation for 'onlineSavingsMsg'
Learn More

6-Cyano-7-nitro-1,4-dihydroquinoxaline-2,3-dione, 98%

Product Code. p-7042712
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470307 10 mg 10mg
15480307 25 mg 25mg
15490307 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15470307 Supplier Thermo Scientific Alfa Aesar Supplier No. J63472.MA
Add to basket
This item is not returnable. View return policy

Potent, selective kainate/quisqualate (non-NMDA) receptor antagonist | CAS: 115066-14-3 | C9H4N4O4 | 232.155 g/mol

Potent, selective kainate/quisqualate (non-NMDA) receptor antagonist

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115066-14-3
Molecular Formula C9H4N4O4
Molecular Weight (g/mol) 232.155
MDL Number MFCD00069232
InChI Key RPXVIAFEQBNEAX-UHFFFAOYSA-N
Synonym cnqx, 6-cyano-7-nitroquinoxaline-2,3-dione, unii-6ote87sccw, 7-nitro-2,3-dioxo-1,2,3,4-tetrahydroquinoxaline-6-carbonitrile, 6ote87sccw, 3h cnqx, 6-cyano-2,3-dihydroxy-7-nitroquinoxaline, 1,4-dihydro-2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile, 6-quinoxalinecarbonitrile, 1,2,3,4-tetrahydro-7-nitro-2,3-dioxo, 2,3-dihydroxy-7-nitro-6-quinoxalinecarbonitrile
PubChem CID 3721046
ChEBI CHEBI:34468
IUPAC Name 7-nitro-2,3-dioxo-1,4-dihydroquinoxaline-6-carbonitrile
SMILES C1=C(C(=CC2=C1NC(=O)C(=O)N2)[N+](=O)[O-])C#N

Specifications

Melting Point >300°C
Color Yellow
Quantity 10 mg
Beilstein 5821072
Solubility Information Soluble in DMSO to 100mM
Formula Weight 232.16
Percent Purity ≥99%
Physical Form Solid
Chemical Name or Material 6-Cyano-7-nitro-1,4-dihydroquinoxaline-2,3-dione

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.