missing translation for 'onlineSavingsMsg'
Learn More

6-Chloronicotinic Acid Ethyl Ester, TRC

Product Code. 30360163
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30360163 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30360163 Supplier Toronto Research Chemicals Supplier No. TRCC37368025G
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Chloronicotinic Acid Ethyl Ester
CAS 49608-01-7
Molecular Formula C8H8ClNO2
Quantity 25 g
Synonym 6-Chloro-3-pyridinecarboxylic Acid Ethyl Ester, 2-Chloropyridine-5-carboxylic Acid Ethyl Ester, Ethyl 2-chloro-5-pyridinecarboxylate, Ethyl 6-Chloro-3-nicotinate, Ethyl 6-Chloro-3-pyridinecarboxylate, Ethyl 6-Chloronicotinate
InChI Formula InChI=1S/C8H8ClNO2/c1-2-12-8(11)6-3-4-7(9)10-5-6/h3-5H,2H2,1H3
SMILES CCOC(=O)c1ccc(Cl)nc1
IUPAC Name ethyl 6-chloropyridine-3-carboxylate
Molecular Weight (g/mol) 185.61
Formula Weight 185.0244
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.