missing translation for 'onlineSavingsMsg'
Learn More

6-Chloroguanine Crystalline MP Biomedicals

Product Code. 11411601 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11411601 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11411601 Supplier MP Biomedicals Supplier No. 0219017901

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 10310-21-1
Molecular Formula C5H4ClN5
MDL Number MFCD00075252
Quantity 1 g
Synonym 2-amino-6-chloropurine, 6-chloroguanine, 6-chloro-9h-purin-2-amine, 6-chloro-2-aminopurine, 2-amino-6-chlorpurine, 1h-purin-2-amine, 6-chloro, purine, 2-amino-6-chloro, 6-chloro-7h-purin-2-ylamine, 2-amino-6-chloro-purin, 2-amino-6-chloro-purine
InChI Key RYYIULNRIVUMTQ-UHFFFAOYSA-N
SMILES NC1=NC(Cl)=C2NC=NC2=N1
IUPAC Name 6-chloro-7H-purin-2-amine
Molecular Weight (g/mol) 169.57
PubChem CID 5360349
ChEBI CHEBI:72345
Formula Weight 169.6
Physical Form Crystalline
Chemical Name or Material 6-Chloroguanine
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.