Learn More
6-Bromopurine, TRC
Specifications
Specifications
| Chemical Name or Material | 6-Bromopurine |
| CAS | 767-69-1 |
| Molecular Formula | C5H3BrN4 |
| Quantity | 10 g |
| Synonym | 6-Bromo-1H-purine, 6-Bromo-9H-purine, 6-Bromopurine, NSC 45150 |
| InChI Formula | InChI=1S/C5H3BrN4/c6-4-3-5(9-1-7-3)10-2-8-4/h1-2H,(H,7,8,9,10) |
| SMILES | Brc1ncnc2nc[nH]c12 |
| IUPAC Name | 6-bromo-7H-purine |
| Molecular Weight (g/mol) | 199.01 |
| Formula Weight | 197.9541 |