Learn More
6-Aminobenzo[d]oxazol-2(3H)-one, 95%, BLD Pharm™
Specifications
Specifications
| CAS | 22876-17-1 |
| Molecular Formula | C7H6N2O2 |
| MDL Number | MFCD00463899 |
| Quantity | 250 mg |
| SMILES | NC1=CC2=C(NC(=O)O2)C=C1 |
| Molecular Weight (g/mol) | 150.13 |
| Percent Purity | 95% |
| Chemical Name or Material | 6-Aminobenzo[d]oxazol-2(3H)-one |