Learn More
6-Amino-5-azacytidine, MedChemExpress
Product Code. 16454508
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16454508 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 16454508 Supplier Medchem Express Supplier No. HY111643
 Restricted Item
Estimated Shipment: 16-11-2026
MedChemExpress 6-Amino-5-azacytidine inhibits the growth of bacteria E. coli.
6-Amino-5-azacytidine inhibits the growth of bacteria E. coli.
Specifications
Chemical Name or Material 6-Amino-5-azacytidine
CAS 105331-00-8
Color White
Purity Grade Notes Research
Molecular Formula C8H13N5O5
Quantity 10 mg
Solubility Information H2O : 14.71 mg/mL (56.75 mM; Need ultrasonic)
SMILES O=C1N([C@H]2[C@H](O)[C@H](O)[C@@H](CO)O2)C(N)=NC(N)=N1
Molecular Weight (g/mol) 259.22
Formula Weight 259.22
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation