Learn More
6-Amino-3-bromo-2-picoline, 98%, Thermo Scientific™
Product Code. 10065113 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
Packaging:
Glass bottle
Unit Size:
5g
Product Code. Quantity Packaging unitSize
10065113 5 g Glass bottle 5g
1 options
Product Code. 10065113
Brand: Thermo Scientific Acros 360750050
Request Bulk or Custom Format
Specifications
CAS 42753-71-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 82°C to 83°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C6H8BrN2
MDL Number MFCD00068230
Quantity 5 g
Synonym 2-amino-5-bromo-6-methylpyridine, 6-amino-3-bromo-2-methylpyridine, 6-amino-3-bromo-2-picoline, 5-bromo-6-methyl-2-pyridinamine, 2-amino-5-bromo-6-picoline, 2-pyridinamine, 5-bromo-6-methyl, 5-bromo-6-methyl-pyridin-2-ylamine, 5-bromo-6-methyl-pyridin-2-amine, 5-bromo-6-methyl-2-pyridylamine, 5-bromo-6-methylpyridin-2-ylamine
InChI Key SEOZHXRTVJPQPZ-UHFFFAOYSA-O
SMILES CC1=[NH+]C(N)=CC=C1Br
IUPAC Name 5-bromo-6-methylpyridin-2-amine
Molecular Weight (g/mol) 188.05
PubChem CID 170678
Formula Weight 187.04
Percent Purity 98%
Physical Form Fluffy Powder
Chemical Name or Material 6-Amino-3-bromo-2-picoline
Show More Show Less
compliance-icons
Product Identifier
  • 6-Amino-3-bromo-2-picoline
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.