missing translation for 'onlineSavingsMsg'
Learn More

6-Amino-1-methyluracil, TRC

Product Code. 30310099
Change view
Click to view available options
Quantity:
100 g
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30310099 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30310099 Supplier Toronto Research Chemicals Supplier No. TRCA616935100G
 Restricted Item
Estimated Shipment: 26-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Amino-1-methyluracil
CAS 2434-53-9
Molecular Formula C5H7N3O2
Quantity 100 g
Synonym 6-Amino-1-methyl-2,4-pyrimidinedione, 1-Methyl-6-aminouracil, 6-Amino-1-methyl-1H-pyrimidine-2,4-dione, 6-Amino-1-methyluracil, NSC 7369
InChI Formula InChI=1S/C5H7N3O2/c1-8-3(6)2-4(9)7-5(8)10/h2H,6H2,1H3,(H,7,9,10)
SMILES CN1C(=O)NC(=O)C=C1N
IUPAC Name 6-amino-1-methylpyrimidine-2,4-dione
Molecular Weight (g/mol) 141.13
Formula Weight 141.0538
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.